Learn More
Selleck Chemical LLC AMI-1 5mg 20324-87-2
SDP
Supplier:  Selleck Chemical LLC S78845MG
Encompass_Preferred
AMI-1 is a potent and specific Histone Methyltransferase (HMT) inhibitor with IC50 of 3.0 ?M and 8.8 ?M for yeast Hmt1p and human PRMT1, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8093