Learn More
AdipoGen Amicarbazone
SDP
Supplier:  AdipoGen CDXA0259M100
Encompass_Preferred
Chemical. CAS 129909-90-6. Formula C10H19N5O2. MW 241.3. Synthetic. Herbicide with a broad spectrum of weed control. Potent inhibitor of photosynthetic electron transport, inducing chlorophyll fluorescence and interrupting oxygen evolution via binding to the Qb domain of photosystem II PSII. Compound can be used as analytical reference material.
Catalog No. 50-256-1777