Learn More
Cayman Chemical AmIcoumacIn B 2.5mg
SDP
Supplier:  Cayman Chemical 317772.5 MG
Encompass_Preferred
An amicoumacin with quorum-sensing inhibitory activity; active against C. violaceum (MIC = 250 UG/ml); increases the expression of BMP2 in MG-63 cells
Catalog No. 50-299-1182