Learn More
Medchemexpress LLC Amidephrine | 37571-84-9 | 97.0% | 244.31 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY136474
Encompass_Preferred
Amidephrine is a selective agonist of alpha 1-adrenoceptor. It inhibits monopulse field contraction of the vas deferens epithelium and prostate gland. This product is for research use only and not for sale to patients.
  • Selective agonist of alpha 1-adrenoceptor
  • Inhibits monopulse field contraction of the vas deferens epithelium and prostate gland
  • For research use only
Catalog No. 50-003-51892