Learn More
Medchemexpress LLC Amido black 10B (naphthol blue black) | 1064-48-8 | MFCD00004017 | 98.2% | 616.49 g/mol | C22H14N6Na2O9S2 | 25g
SDP
Supplier:  Medchemexpress LLC HYY069525G
Encompass_Preferred
Amido Black 10B (Naphthol Blue Black) is a highly toxic azo dye Amido Black 10B is genotoxic and mutagenic Amido Black 10B can cause respiratory problems Amido Black 10B is used for amino acid dyeing[1][2][3][4]
Catalog No. 50-002-69858