Learn More
Medchemexpress LLC Amidosulfuron | 120923-37-7 | MFCD01310639 | 98.4% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB191250MG
Encompass_Preferred
Amidosulfuron is a herbicide that targets acetohydroxyacid synthase (AHAS) and is highly effective against dicot broad-leaf weeds. It is for research use only.
  • Targets acetohydroxyacid synthase (AHAS)
  • Highly effective against dicot broad-leaf weeds
  • Reduces photosynthetic rate in treated plants
  • Reduces leaf chlorophyll, soluble sugar and pyruvate content
  • Alters content of several free amino acids
  • Appears as a white to off-white solid
  • Soluble in DMSO
Catalog No. 50-004-12146