Learn More
Medchemexpress LLC Amifampridine (3,4-Diaminopyridine) | 54-96-6 | MFCD00006401 | >99.9% | 109.13 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY14946
Encompass_Preferred
Amifampridine (3,4-Diaminopyridine) is an orally active, potent, and cell-permeable voltage-gated potassium (Kv) channel blocker (PCB). It is effective in reversing BoNT/A intoxication and increases transmitter release from neuromuscular junctions (NMJs). Amifampridine can be used for Lambert-Eaton myasthenic syndrome (LEMS) research.
  • Orally active, potent, and cell-permeable voltage-gated potassium (Kv) channel blocker.
  • Effective in reversing BoNT/A intoxication.
  • Increases transmitter release from neuromuscular junctions (NMJs).
  • Used for research related to Lambert-Eaton myasthenic syndrome (LEMS).
Catalog No. 50-003-63256