Learn More
Abcam Amifostine, cytoprotective agent, 10MG
Supplier:  Abcam AB14106010MG
Encompass_Preferred
MW 214.23 Da, Purity >98%. Broad spectrum cytoprotective agent. Inactive prodrug of WR-1065. In vitro effects include p53 activation and indirect activation of proteins, including Bcl-2 and HIF-1α, involved in DNA repair and apoptosis. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-5995