Learn More
Medchemexpress LLC Amifostine (trihydrate) | 112901-68-5 | 100.0% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0639A200MG
Encompass_Preferred
Amifostine trihydrate is a broad-spectrum cytoprotective agent and radioprotector. It selectively protects normal tissues from damage caused by radiation and chemotherapy. It acts as a potent hypoxia-inducible factor-α1 (HIF-α1) and p53 inducer, protecting cells by scavenging oxygen-derived free radicals. It also reduces renal toxicity and exhibits antiangiogenic action.
  • Broad-spectrum cytoprotective agent
  • Radioprotector
  • Selectively protects normal tissues from radiation and chemotherapy damage
  • Potent HIF-α1 and p53 inducer
  • Scavenges oxygen-derived free radicals
  • Reduces renal toxicity
  • Exhibits antiangiogenic action
  • Protects against tert-Butyl hydroperoxide (TBHP)-induced cell damage
  • Reduces H9c2 cell apoptosis
  • Provides protection against myocardial ischemia/reperfusion injury
Catalog No. 50-004-08512