Learn More
Medchemexpress LLC Amifostine trihydrate | 112901-68-5 | 99.95% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0639A5MG
Encompass_Preferred
Amifostine trihydrate is a broad-spectrum cytoprotective agent and radioprotector. It selectively protects normal tissues from damage caused by radiation and chemotherapy. This agent acts as a potent hypoxia-inducible factor-α1 (HIF-α1) and p53 inducer, protecting cells by scavenging oxygen-derived free radicals, reducing renal toxicity, and exhibiting antiangiogenic action.
  • Selectively protects normal tissues from radiation and chemotherapy damage.
  • Potent hypoxia-inducible factor-α1 (HIF-α1) and p53 inducer.
  • Protects cells by scavenging oxygen-derived free radicals.
  • Reduces renal toxicity.
  • Exhibits antiangiogenic action.
  • Reduces tert-butyl hydroperoxide (TBHP)-induced cell damage in vitro.
  • Attenuates cardiomyocyte apoptosis and reduces reactive oxygen species (ROS) in vivo.
Catalog No. 50-004-08557