Learn More
Selleck Chemical LLC Amikacin E4932-50MG
SDP
Supplier:  Selleck Chemical LLC E493250MG
Encompass_Preferred
Amikacin(BAY 41-6551 Lukadin) is a semisynthetic aminoglycoside derived from kanamycin-A that acts as an inhibitor of the Z ring formation leading to the inhibition of cell division It is used to treat serious infections caused by aerobic Gram-negative bacteria mycobacteria Nocardia and life-threatening infections in neonates
Catalog No. 50-000-43331