Learn More
Medchemexpress LLC Amikacin hydrate | 1257517-67-1 | MFCD11045969 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0509200MG
Encompass_Preferred
Amikacin hydrate is an aminoglycoside antibiotic and a semisynthetic analog of kanamycin. It is bactericidal, inhibiting protein synthesis by acting directly on the 30S and 50S bacterial ribosomal subunits.
  • Highly active against most Gram-negative bacteria
  • Effective against gentamicin- and tobramycin-resistant strains
  • Inhibits infections caused by susceptible Nocardia and nontuberculous mycobacteria
  • Treats infections caused by organisms resistant to other aminoglycosides
  • Less affected by aminoglycoside-modifying enzymes
Catalog No. 50-004-06841