Learn More
Medchemexpress LLC Amikacin (standard) | 37517-28-5 | 99.9% | 200 MG

Supplier: Medchemexpress LLC HYB0509A200MG
Amikacin is a semisynthetic kanamycin analog that is effective against most Gram-negative bacteria, including strains resistant to gentamicin and tobramycin. It exhibits a significant inhibitory effect and can be utilized in bacteriostatic, anti-cancer, and analgesic studies.
- Semisynthetic kanamycin analog
- Effective against most Gram-negative bacteria
- Exhibits significant inhibitory effect
- Can be utilized in bacteriostatic, anti-cancer, and analgesic studies
- Appearance: solid, white to off-white
- Purity: 99.9%
- Molecular weight: 585.60
- Formula: C22H43N5O13
- Recommended storage: 4°C, sealed and dry, away from moisture
- Shipping: room temperature in continental US
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.