Learn More
Selleck Chemical LLC Amiloride HCl 50mg 2016-88-8
SDP
Supplier:  Selleck Chemical LLC S181150MG
Encompass_Preferred
Amiloride is a selective T-type calcium channel blocker, an epithelial sodium channel blocker and a selective inhibitor of urokinase plasminogen activator (uPA)(Ki=7 ?M). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1656