Learn More
Enzo Life Sciences Amiloride hydrochloride dihydrate (1g). CAS: 17440-83-4
SDP
Supplier:  Enzo Life Sciences ALX550212G001
Encompass_Preferred
Selectively blocks low threshold (T-type) calcium channel (KD=30μM) with no effect on L-type channels or tetrodotoxin Na+ channels. Inhibits the epithelial tetrodotoxin insensitive Na+ channel (IC50 =1μM). Clinically useful K+ sparing diuretic. Alternative name: 3 5-Diamino-N-(aminoiminomethyl)-6-chloropyrazinecarboxamide HCl 2H2O. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or water (1mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-200-9498