Learn More
Medchemexpress LLC Amiloride hydrochloride | 2016-88-8 | MFCD00211292 | 99.8% | 266.09 g/mol | C6H9Cl2N7O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0285AR10MG
Encompass_Preferred
Amiloride hydrochloride is the analytical reference standard of amiloride intended for research and analytical applications. It functions as an inhibitor of epithelial sodium channels (ENaC) and urokinase-type plasminogen activator receptor (uPA), and blocks polycystin-2 (PC2/TRPP2) channels.
  • High purity analytical standard (99.8%).
  • Suitable for assay development, calibration, and method validation.
  • Available in small reference quantities for laboratory use, such as 10 mg.
  • Solid powder form, stable under recommended storage conditions.
Catalog No. 50-002-98271