Learn More
Medchemexpress LLC Amine-PEG-CH2COOH (MW 3400) | 99.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140644100MG
Encompass_Preferred
Amine-PEG-CH2COOH (MW 3400) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-53866