Learn More
Medchemexpress LLC Amine-PEG3-Desthiobiotin | 2237234-71-6 | 99.9% | 388.50 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY134721
Encompass_Preferred
Amine-PEG3-Desthiobiotin is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-66085