Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amine-PEG6-thiol hydrochloride | 00-00-0 | 377.92 g/mol | C14H32ClNO6S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY134711A5MG

Encompass_Preferred

Amine-PEG6-thiol hydrochloride is a PEG-based bifunctional linker (hydrochloride salt) designed for use in PROTAC and conjugation chemistry. It provides orthogonal reactive handles that enable flexible attachment strategies and improves solubility and linker flexibility for bifunctional degrader construction. The reagent is supplied as a viscous liquid with defined storage recommendations to maintain stability.

  • Provides amine and thiol functional handles for conjugation.
  • PEG6 spacer increases solubility and conformational flexibility.
  • Offered as the hydrochloride salt to enhance stability.
  • Viscous liquid form compatible with solution-phase coupling workflows.
  • Storage recommendations: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Molecular weight 377.92 g/mol and formula C14H32ClNO6S.

Catalog No. 50-002-23395


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.