Learn More
Biotium Amine Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF410 1/EA
Supplier:  Biotium 97502
Encompass_Preferred
CF Dye Succinimidyl Esters (SE also known as NHS esters) are Biotiums next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups such as lysine residues Blue fluorescent CF410 has an excitation/emission at 404/455 nm
Catalog No. 50-005-01463