Learn More
Hach Company Amino Acid Reagent, 500 mL
Supplier:  Hach Company 193449
Encompass
Reagent for Phosphate determination by the Amino Acid method. 500 mL bottle. Method Name: Amino Acid.
Catalog No. 50-465-239