Learn More
Medchemexpress LLC Amino-PEG10-OH | 129449-09-8 | MFCD28385478 | >98.0% | 457.56 g/mol | C20H43NO10 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY120761250MG
Encompass_Preferred
Amino-PEG10-OH is a 10-unit polyethylene glycol (PEG) linker bearing a terminal primary amine, used as a non-cleavable spacer in bioconjugation. It is commonly employed in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs) to provide hydrophilic spacing and a reactive site for payload attachment. The material is typically supplied as a white to off-white solid (CAS 129449-09-8) with a molecular weight of 457.56 g/mol.
  • Non-cleavable 10-unit PEG spacer suitable for ADC and PROTAC chemistry.
  • Terminal primary amine enables straightforward conjugation to activated esters and acids.
  • Hydrophilic spacer improves solubility and reduces steric hindrance between conjugated components.
  • High purity suitable for research applications.
  • Available in laboratory quantities such as 250 mg for small-scale synthesis.
Catalog No. 50-002-03455