Learn More
Medchemexpress LLC Amino-PEG12-alcohol | 1345681-71-1 | MFCD27083070 | ≥98% | C25H53NO13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140205100MG
Encompass_Preferred
Amino-PEG12-alcohol is a PEG linker that is used in the synthesis of ADCs (antibody-drug conjugates) and other conjugates.
  • Improves the aqueous solubility of ADCs
Catalog No. 50-003-57234