Learn More
Medchemexpress LLC Amino-PEG16-acid | 2922656-52-6 | 95.0% | C35H71NO18 | 25MG
Supplier: Medchemexpress LLC 25MGHY140179
Amino-PEG16-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are molecules that utilize a linker to connect two ligands: one for an E3 ubiquitin ligase and another for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
- PEG-based linker for PROTAC synthesis.
- Enables selective degradation of target proteins.
- Utilizes the intracellular ubiquitin-proteasome system.
- Solid appearance, white to off-white color.
- For research use only.