Learn More
Medchemexpress LLC Amino-PEG16-acid | 2922656-52-6 | MFCD26793767 | 95.0% | 793.93 | C35H71NO18 | 5 MG

Supplier: Medchemexpress LLC HY1401795MG
Amino-PEG16-acid is a polyethylene glycol (PEG)-based linker bearing a terminal amine and carboxylic acid. It provides a flexible, water-soluble spacer for PROTAC synthesis and bioconjugation, supplied as a solid with reported purity of 95.0% and molecular weight of 793.93 g/mol. Analytical documents and handling instructions (SDS, COA, HNMR, MS) are available from the manufacturer.
- Provides a flexible, hydrophilic spacer for linking ligands.
- Contains reactive amine and carboxylic acid functional groups for conjugation.
- Suitable for PROTAC synthesis and other bioconjugation workflows.
- Supplied as a solid with reported purity of 95.0%.
- Soluble in common polar solvents such as DMSO, DMF, and methanol.
- Powder stable at -20°C for long-term storage; specific in-solution stability limited to refrigerated/ultra-low temperatures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.