Learn More
Apexbio Technology LLC Amino-PEG2-C2-acid 791028-27-8 5g
SDP
Supplier:  Apexbio Technology LLC BA24445000
Encompass_Preferred
Amino-PEG2-C2-acid (CAS 791028-27-8) is a bifunctional polyethylene glycol (PEG)-based linker featuring both amino and carboxyl functional groups It is primarily utilized in the synthesis of antibody-drug conjugates (ADCs) facilitating the covalent attachment of active pharmaceutical molecules to antibodies By introducing a short hydrophilic PEG spacer Amino-PEG2-C2-acid can enhance solubility and modulate the pharmacokinetic properties of conjugated compounds The compound s dual reactivity enables efficient conjugation strategies in biomedical research and drug development
Catalog No. 50-000-89471