Learn More
Medchemexpress LLC Amino-PEG4-alcohol | 86770-74-3 | 99.8% | 193.24 g/mol | C8H19NO4 | 5 G

Supplier: Medchemexpress LLC HYW0080055G
Amino-PEG4-alcohol is an amino-terminated four-unit polyethylene glycol (PEG4) linker with a primary amine and a terminal hydroxyl. It functions as a hydrophilic, bifunctional spacer for conjugation chemistries, commonly used in PROTAC and ADC synthesis, and for introducing PEG spacing between ligands or payloads.
- Contains a primary amine and a terminal hydroxyl.
- Four-unit PEG spacer provides hydrophilicity and flexibility.
- Suitable for PROTAC and ADC linker synthesis.
- Molecular weight approximately 193.24 g/mol and CAS 86770-74-3.
- Available as a 5 g package for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.