Learn More
Medchemexpress LLC Amino-PEG4-C1-Boc | 864680-64-8 | 307.38 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY42619
Encompass_Preferred
Amino-PEG4-C1-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Detailed solvent and solubility data available for in vivo applications (PBS, DMSO, PEG300, SBE-β-CD in saline, corn oil).
  • Functions as a PROTAC linker, connecting ligands for E3 ubiquitin ligase and target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system for selective protein degradation.
Catalog No. 50-004-02815