Learn More
Medchemexpress LLC Amino-PEG4-C2-SH (hydrochloride) | 2762393-21-3 | >95.0% | 289.82 | 50 MG

Supplier: Medchemexpress LLC HY13844650MG
Thiol-PEG4-NH2 hydrochloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This linker is crucial as PROTACs contain two different ligands connected by such a linker; one targets an E3 ubiquitin ligase and the other targets the protein of interest. PROTACs leverage the intracellular ubiquitin-proteeasome system to selectively degrade target proteins.
- Peg-based protac linker.
- Can be used in the synthesis of protacs.
- Appearance: liquid.
- Color: colorless to light yellow.
- Shipping at room temperature in continental US; may vary elsewhere.
- Storage at -20°C, protect from light, stored under nitrogen.
- For solvent solutions: store at -80°C for 6 months or -20°C for 1 month (protect from light, stored under nitrogen).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.