Learn More
Medchemexpress LLC Amino-PEG4-CH2COOH | 195071-49-9 | ≥98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1305245G
Encompass_Preferred
Amino-PEG4-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It also serves as a non-cleavable 4-unit PEG ADC linker for the creation of antibody-drug conjugates (ADCs).
  • Used in the synthesis of PROTACs
  • Functions as a non-cleavable 4-unit PEG ADC linker
  • PROTACs degrade target proteins using the ubiquitin-proteasome system
  • ADCs consist of an antibody linked to an ADC cytotoxin
Catalog No. 50-003-38151