Learn More
Medchemexpress LLC Amino-PEG4-GGFG-Dxd | 2879227-88-8 | 1088.14 g·mol⁻1 | C53H66FN9O15 | 5 MG

Supplier: Medchemexpress LLC HY1587685MG
Amino-PEG4-GGFG-Dxd is a drug-linker conjugate intended for use in antibody-drug conjugate (ADC) synthesis. It couples a DXd (deruxtecan) payload to a GGFG peptide-based linker via a PEG4 spacer and provides an amino functional handle for conjugation to antibodies.
- drug-linker conjugate for ADC synthesis
- contains a DXd payload linked via a cleavable GGFG peptide linker
- includes a PEG4 spacer to improve solubility and spacing
- features an amino functional handle for conjugation chemistry
- available in small milligram quantities for research use
- molecular weight 1088.14 g·mol⁻1, formula C53H66FN9O15
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.