Learn More
Medchemexpress LLC Amino-PEG4-GGFG-Dxd | 2879227-88-8 | MFCD09950912 | 100.0% | 1088.14 | C53H66FN9O15 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1587681MG
Encompass_Preferred
Amino-PEG4-GGFG-Dxd is a drug-linker conjugate designed for the synthesis of antibody-drug conjugates (ADCs). It consists of the DXd cytotoxic payload connected via a PEG4-GGFG peptide linker with a terminal amino group, and is supplied as an off-white to light yellow solid for research use only.
  • Drug-linker conjugate intended for ADC synthesis.
  • Contains DXd (an exatecan derivative) as the cytotoxic payload.
  • PEG4-GGFG peptide linker provides spacer and protease-cleavable sequence.
  • Terminal amino group enables conjugation to activated linkers or antibodies.
  • High stated purity and solid form suitable for research applications.
Catalog No. 50-002-04442