Learn More
Medchemexpress LLC Amino-PEG4-Val-Cit-PAB-MMAE | 1492056-71-9 | 99.9% | 1370.71 g/mol | C69H115N11O17 | 5 MG

Supplier: Medchemexpress LLC HY1411545MG
Amino-PEG4-Val-Cit-PAB-MMAE is a cleavable four-unit PEG linker terminating in an amino group and bearing a Val-Cit-PAB cleavable spacer attached to monomethyl auristatin E (MMAE). It is supplied for research use and is used in the synthesis of antibody-drug conjugates (ADCs).
- Cleavable Val-Cit-PAB spacer linking to MMAE.
- Four-unit PEG spacer with amino terminal.
- High purity: 99.93% (manufacturer reported).
- Molecular weight approximately 1370.71 g/mol.
- Chemical formula: C69H115N11O17.
- Soluble in DMSO; in-vivo formulation guidance provided by manufacturer.
- Store sealed at -20°C; unstable in solution-prepare fresh solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.