Learn More
Medchemexpress LLC Amino-PEG5-amine | 72236-26-1 | 97.0% | 10 G

Supplier: Medchemexpress LLC HY13044710G
Amino-PEG5-amine is a PEG-based (5 units) PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only.
- PROTAC linker
- PEG-based (5 units)
- Used in the synthesis of PROTACs
- Appears as a colorless to light yellow liquid
- Molecular weight: 280.36
- Soluble in water (≥ 100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.