Learn More
Medchemexpress LLC Amino-PEG5-Boc | 1446282-18-3 | 98.0% | C17H35NO7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140194
Encompass_Preferred
Amino-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system for degradation
  • Enables selective degradation of target proteins
Catalog No. 50-003-52828