Learn More
Medchemexpress LLC Amino-PEG5-C2-acid | 309.36 g·mol⁻1 | C13H27NO7 | 5G
SDP
Supplier:  Medchemexpress LLC HY1153845G
Encompass_Preferred
Amino-PEG5-C2-acid is a PEG-based PROTAC linker can be used in the synthesis of PROTACs Amino-PEG5-C2-acid is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1]
Catalog No. 50-002-28156