Learn More
Medchemexpress LLC Amino-PEG5-CH2COOH | 141282-35-1 | ≥97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130577100MG
Encompass_Preferred
Amino-PEG5-CH2COOH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Peg-based protac linker
  • Utilized in the synthesis of protacs
  • Consists of two distinct ligands connected by a linker
  • Targets E3 ubiquitin ligase and protein of interest
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-38427