Learn More
Medchemexpress LLC Amino-PEG5-CH2COOH | 141282-35-1 | >=97.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13057750MG
Encompass_Preferred
Amino-PEG5-CH2COOH is a PEG-based PROTAC linker essential for the synthesis of PROTACs. PROTACs are innovative molecules that feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This design allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables PROTACs to exploit the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-40184