Learn More
Medchemexpress LLC Amino-PEG6-amido-bis-PEG5-N3 | 1147.31 | C48H94N10O21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1309575MG
Encompass_Preferred
Amino-PEG6-amido-bis-PEG5-N3 is a cleavable 11-unit polyethylene glycol (PEG) linker bearing amino and azide functional groups, designed for use in antibody-drug conjugate synthesis and bioconjugation via click chemistry.
  • Cleavable 11-unit PEG linker useful for ADC construction.
  • Contains an azide group for copper-catalyzed and strain-promoted click reactions.
  • Amino terminus enables conjugation or further functionalization.
  • Improves solubility and flexibility between linked molecules.
  • Available in small research quantities for conjugation workflows.
Catalog No. 50-002-03234