Learn More
Medchemexpress LLC Amino-PEG6-amido-bis-PEG5-N3 | 1147.31 g/mol | C48H94N10O21 | 100 MG

Supplier: Medchemexpress LLC HY130957100MG
Amino-PEG6-amido-bis-PEG5-N3 is a cleavable PEG-based linker designed for bioconjugation and click chemistry applications. It contains an azide functional group and an amino terminus, providing an azide handle suitable for copper-catalyzed azide-alkyne cycloaddition and formation of antibody-drug conjugates or other bioconjugates.
- Cleavable 11-unit PEG linker for ADC and bioconjugate synthesis.
- Azide functional group compatible with CuAAC (click chemistry).
- Molecular weight 1,147.31 g/mol; formula C48H94N10O21.
- Appearance oil; colorless to light yellow.
- Solubility in DMSO ≥ 100 mg/mL.
- Storage sealed, away from moisture and light; recommended -20°C (in solvent: -80°C, 6 months; -20°C, 1 month).
- Available in multiple pack sizes including 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.