Learn More
Medchemexpress LLC Amino-PEG7-C2-SH hydrochloride | 00-00-0 | 98.0% | C16H36ClNO7S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13845110MG
Encompass_Preferred
Amino-PEG7-C2-SH hydrochloride is a PEG-based PROTAC linker supplied as the hydrochloride salt. It provides a PEG7 spacer with a terminal amino group and a short C2 thiol handle for conjugation in PROTAC synthesis. The material is a light yellow liquid with a molecular weight of 421.98 g/mol and reported purity of 98.0%. Store sealed at -20°C; in solution, store at -80°C for long-term stability.
  • Provides a PEG7 spacer for flexible linker design.
  • Terminal amino group enables amide or carbamate coupling.
  • C2 thiol handle allows disulfide or thioether conjugation.
  • High purity (98.0%) suitable for research synthesis.
  • Liquid form simplifies handling in small-scale reactions.
  • Stable when stored sealed at recommended temperatures.
Catalog No. 50-002-60997