Learn More
TARGETMOL CHEMICALS INC Amino-PEG8-amine 250MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T14250250MG
Encompass_Preferred
Also available in 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Amino-PEG8-amine is a pegylated based PROTAC linker that can be used in the synthesis of PROTACs. It is a conjugate PEG derivative containing two amino (NH2) groups. Purity 98.27%
Catalog No. 50-287-7007