Learn More
Medchemexpress LLC Amino-PEG8-Boc | 756526-06-4 | 99.8% | 497.62 g/mol | C23H47NO10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW019799100MG
Encompass_Preferred
Amino-PEG8-Boc is a Boc-protected amino polyethylene glycol linker composed of eight ethylene glycol units. It is used as a cleavable spacer in antibody-drug conjugates (ADCs) and as a PEG-based linker in PROTAC synthesis, providing hydrophilicity and a reactive amine for further functionalization.
  • Cleavable 8-unit PEG linker.
  • Boc-protected amino terminus for selective deprotection and functionalization.
  • Improves solubility and provides spacing between conjugated moieties.
  • High purity (≈99.8%).
  • Available in 100 mg packaging.
  • Storage: pure form -20°C (long term) or 4°C (short term).
Catalog No. 50-002-03932