Learn More
Medchemexpress LLC Amino-PEG9-acid | 1191079-83-0 | MFCD26127809 | 98.0% | 485.57 g/mol | C21H43NO11 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY130166250MG
Encompass_Preferred
Amino-PEG9-acid is a polyethylene glycol (PEG)-based linker used in the synthesis of proteolysis targeting chimeras (PROTACs) and as a non-cleavable linker for antibody-drug conjugates (ADCs). It is typically supplied as a solid with high purity for use in linker chemistry and bioconjugation applications.
  • PEG-based linker with an amino terminus and a terminal carboxylic acid.
  • Suitable for PROTAC and ADC synthesis and other bioconjugation reactions.
  • High purity, suitable for research and synthetic chemistry applications.
  • Soluble in common organic solvents and aqueous mixtures for coupling reactions.
  • Available in multiple package sizes to support small-scale and larger syntheses.
Catalog No. 50-002-02465