Learn More
Medchemexpress LLC Amino-PEG9-alcohol | 15332-95-3 | 98.0% | C18H39NO9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140204100MG
Encompass_Preferred
Amino-PEG9-alcohol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs.
  • Enables the connection of two different ligands within PROTACs
  • Facilitates PROTACs to exploit the intracellular ubiquitin-proteasome system
  • Supports selective degradation of target proteins
Catalog No. 50-003-56757