Learn More
Medchemexpress LLC Amino-PEG9-alcohol | 15332-95-3 | 98.0% | C18H39NO9 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140204
Encompass_Preferred
Amino-PEG9-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by containing two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-52510