Learn More
Medchemexpress LLC Amino-PEG9-amine | 474082-35-4 | >98% | C20H44N2O9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140210100MG
Encompass_Preferred
Amino-PEG9-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56798