Learn More
Medchemexpress LLC Amino Tadalafil 100mg | 385769-84-6 | 100 MG

Supplier: Medchemexpress LLC HY117109100MG
Amino tadalafil is a structural analogue of tadalafil provided as a research chemical. It acts as a phosphodiesterase 5 (PDE5) inhibitor and is intended for research and analytical applications only; not for human or therapeutic use.
- High purity (99.04%) suitable for analytical applications.
- Solid, white to off-white powder for ease of handling.
- Molecular formula C21H18N4O4 and molecular weight 390.39 g/mol for accurate formulation.
- Identified by CAS number 385769-84-6 for unambiguous substance matching.
- Recommended storage: store at -20°C and protect from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.
- Available in a 100 MG solid package and in various smaller/larger quantities and DMSO solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.