Learn More
Medchemexpress LLC Amino-tri-(carboxyethoxymethyl)-methane | 174362-95-9 | MFCD31615450 | ≥97.0% | 337.32 g/mol | C13H23NO9 | 250 MG

Supplier: Medchemexpress LLC HY117519250MG
Amino-tri-(carboxyethoxymethyl)-methane is a viscous small-molecule linker reagent (CAS 174362-95-9) used in linker chemistry and specialty organic synthesis. It is a multifunctional tri-carboxyethoxymethyl-bearing amino compound with formula C13H23NO9 and a molecular weight of 337.32 g/mol, supplied in small-scale quantities for research and development applications.
- Viscous liquid form suitable for solution-phase chemistry.
- Contains three carboxyethoxymethyl groups for multifunctional conjugation.
- Chemical formula C13H23NO9 and molecular weight 337.32 g/mol.
- Intended for research use in linker assembly and ADC development.
- Available in small pack sizes to support synthesis and method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.