Learn More
Medchemexpress LLC Aminoacetone hydrochloride | 7737-17-9 | MFCD00056125 | 109.55 | 5 G
Supplier: Medchemexpress LLC HYW0048505G
Aminoacetone hydrochloride is the simplest monopeptide. It is an intermediate in the metabolism of threonine and glycine. It serves as an endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO) and can be used for the determination of SSAO activity.
- Simplest monopeptide
- Intermediate in threonine and glycine metabolism
- Endogenous substrate for semicarbazide-sensitive amine oxidase (SSAO)
- Used for determining SSAO activity